4-Bromo-2-(trifluoromethyl)benzaldehyde

Supplier: Apollo Scientific

PC49241-1G PC49241-25G PC49241-5G
APOSPC49241-1GEA 108 EUR
APOSPC49241-1G APOSPC49241-25G APOSPC49241-5G
4-Bromo-2-(trifluoromethyl)benzaldehyde
4-Bromo-2-(trifluoromethyl)benzaldehyde
Formula: C₈H₄BrF₃O
Melting Pt: 47…50 °C
Storage Temperature: Ambient
MDL Number: MFCD09037472
CAS Number: 861928-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR