Pentafluoroiodobenzene 99%

Supplier: Apollo Scientific
Warning

Synonyms: Iodopentafluorobenzene

PC4930-100G PC4930-5G
APOSPC4930-100GEA 377 EUR
APOSPC4930-100G APOSPC4930-5G
Pentafluoroiodobenzene 99%
Pentafluoroiodobenzene
Formula: C₆F₅I
MW: 293.96 g/mol
Boiling Pt: 166 °C
Melting Pt: -29
Density: 2.204 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001032
CAS Number: 827-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR