Isopropyl trifluoroacetate 99%
Supplier: Apollo Scientific
|
Danger
|
PC4954-25G
PC4954-100G
PC4954-500G
APOSPC4954-25GEA
27.7
EUR
APOSPC4954-25G
APOSPC4954-100G
APOSPC4954-500G
Isopropyl trifluoroacetate 99%
Isopropyl trifluoroacetate
Formula:
C₅H₇F₃O₂ Boiling Pt: 73 °C/749mmHg Melting Pt: < –78 °C Density: 1,108 g/cm³ (25 °C) Flash Pt: –8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013566 CAS Number: 400-38-4 EINECS: 206-922-1 UN: 2924 ADR: 3+8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...