2,2-Difluorocyclopropanemethanol

Supplier: Apollo Scientific
Warning

PC49666-1G PC49666-5G PC49666-250MG
APOSPC49666-1GEA 302 EUR
APOSPC49666-1G APOSPC49666-5G APOSPC49666-250MG
2,2-Difluorocyclopropanemethanol
2,2-Difluorocyclopropanemethanol
Formula: C₄H₆F₂O
MW: 108,09 g/mol
Boiling Pt: 134…135 °C
Storage Temperature: Ambient
MDL Number: MFCD06797748
CAS Number: 509072-57-5
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR