3-Fluoro-5-methylbenzaldehyde, Technical Grade

Supplier: Apollo Scientific

Synonyms: 3-Fluoro-5-tolualdehyde, 5-Fluoro-m-tolylaldehyde, 5-Fluoro-m-tolualdehyde

PC49722-1G PC49722-25G PC49722-5G
APOSPC49722-1GEA 21.9 EUR
APOSPC49722-1G APOSPC49722-25G APOSPC49722-5G
3-Fluoro-5-methylbenzaldehyde, Technical Grade
3-Fluoro-5-methylbenzaldehyde
Formula: C₈H₇FO
MW: 138,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094315
CAS Number: 189628-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR