2-Bromo-5-(trifluoromethyl)phenol
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-Bromo-5-trifluoromethylphenol
PC499002-25G
APOSPC499002-25GEA
111
EUR
APOSPC499002-25G
2-Bromo-5-(trifluoromethyl)phenol
2-Bromo-5-(trifluoromethyl)phenol
Formula:
C₇H₄BrF₃O MW: 241,01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09261190 CAS Number: 402-05-1 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...