(3,5-Difluorophenyl)methanesulphonyl chloride 97%
Supplier: Apollo Scientific
|
Danger
|
PC50011-1G
PC50011-250MG
APOSPC50011-1GEA
87
EUR
APOSPC50011-1G
APOSPC50011-250MG
(3,5-Difluorophenyl)methanesulphonyl chloride 97%
(3,5-Difluorophenyl)methanesulphonyl chloride
Formula:
C₇H₅ClF₂O₂S MW: 226,63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09743349 CAS Number: 163295-74-7 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...