4-(Trifluoromethoxy)phenylmethanesulphonyl chloride
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
4-(Trifluoromethoxy)benzylsulphonyl chloride
PC50014-1G
PC50014-250MG
APOSPC50014-1GEA
109
EUR
APOSPC50014-1G
APOSPC50014-250MG
4-(Trifluoromethoxy)phenylmethanesulphonyl chloride
4-(Trifluoromethoxy)phenylmethanesulphonyl chloride
Formula:
C₈H₆ClF₃O₃S MW: 274.65 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09811274 CAS Number: 683813-55-0 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...