3-Fluoro-5-methoxyphenol

Supplier: Apollo Scientific
Warning

PC500163-10G PC500163-1G PC500163-5G
APOSPC500163-10GEA 376 EUR
APOSPC500163-10G APOSPC500163-1G APOSPC500163-5G
3-Fluoro-5-methoxyphenol
3-Fluoro-5-methoxyphenol
Formula: C₇H₇FO₂
MW: 142,1300 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11848782
CAS Number: 850793-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR