3-Fluoro-2-methyl-6-nitroaniline
Supplier: Apollo Scientific
|
Warning
|
PC500332-250MG
PC500332-5G
APOSPC500332-250MGEA
37.9
EUR
APOSPC500332-250MG
APOSPC500332-5G
3-Fluoro-2-methyl-6-nitroaniline
3-Fluoro-2-methyl-6-nitroaniline
Formula:
C₇H₇FN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD03787364 CAS Number: 485832-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...