2-(Bromomethyl)-4-fluoro-1-nitrobenzene

Supplier: Apollo Scientific
Danger

Synonyms: 5-Fluoro-2-nitrobenzyl bromide

PC500377-1G PC500377-5G
APOSPC500377-1GEA 435 EUR
APOSPC500377-1G APOSPC500377-5G
2-(Bromomethyl)-4-fluoro-1-nitrobenzene
2-(Bromomethyl)-4-fluoro-1-nitrobenzene
Formula: C₇H₅BrFNO₂
MW: 234,0200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09736331
CAS Number: 82420-35-7
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR