3-Fluoro-5-iodobenzotrifluoride

Supplier: Apollo Scientific

Synonyms: 1-Fluoro-3-iodo-5-(trifluoromethyl)benzene

PC500391-100MG PC500391-1G PC500391-250MG
APOSPC500391-100MGEA 178 EUR
APOSPC500391-100MG APOSPC500391-1G APOSPC500391-250MG
3-Fluoro-5-iodobenzotrifluoride
3-Fluoro-5-iodobenzotrifluoride
Formula: C₇H₃F₄I
Storage Temperature: Ambient
MDL Number: MFCD09800718
CAS Number: 1027513-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR