1-Bromo-3-fluoro-5-iodobenzene

Supplier: Apollo Scientific
Warning

Synonyms: 3-Fluoro-5-iodobromobenzene

PC500481-1G PC500481-5G
APOSPC500481-1GEA 66.9 EUR
APOSPC500481-1G APOSPC500481-5G
1-Bromo-3-fluoro-5-iodobenzene
1-Bromo-3-fluoro-5-iodobenzene
Formula: C₆H₃BrFI
MW: 300,9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06657969
CAS Number: 845866-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR