2-Fluoro-5-nitrophenol 98%
Supplier: Apollo Scientific
|
Warning
|
PC5009-25G
PC5009-5G
PC5009-1G
APOSPC5009-25GEA
168
EUR
APOSPC5009-25G
APOSPC5009-5G
APOSPC5009-1G
2-Fluoro-5-nitrophenol 98%
2-Fluoro-5-nitrophenol
Formula:
C₆H₄FNO₃ MW: 157.1000 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09054718 CAS Number: 22510-08-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...