2-Fluoro-3-nitrobenzaldehyde

Supplier: Apollo Scientific
Warning

PC501176-250MG
APOSPC501176-250MGEA 36.4 EUR
APOSPC501176-250MG
2-Fluoro-3-nitrobenzaldehyde
2-Fluoro-3-nitrobenzaldehyde
Formula: C₇H₄FNO₃
MW: 169,1100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08669885
CAS Number: 96516-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR