Magnesium trifluoromethanesulphonate 98%

Supplier: Apollo Scientific
Danger

Synonyms: Magnesium trifluoromethanesulphonate, Magnesium triflate

PC5026-10G PC5026-100G PC5026-25G
APOSPC5026-10GEA 102 EUR
APOSPC5026-10G APOSPC5026-100G APOSPC5026-25G
Magnesium trifluoromethanesulphonate 98%
Magnesium trifluoromethanesulphonate
Formula: C₂F₆MgO₆S₂
MW: 322.45 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00042601
CAS Number: 60871-83-2
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR