2-Fluoro-6-nitroanisole

Supplier: Apollo Scientific
Warning

PC5084-1G PC5084-5G
APOSPC5084-1GEA 27.7 EUR
APOSPC5084-1G APOSPC5084-5G
2-Fluoro-6-nitroanisole
2-Fluoro-6-nitroanisole
Formula: C₇H₆FNO₃
MW: 171,1300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06656241
CAS Number: 484-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR