4-(2,2,2-Trifluoroethylcarbamoyl)phenylboronic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: (4-((2,2,2-Trifluoroethyl)carbamoyl)phenyl)boronic acid

PC5116-1G PC5116-5G
APOSPC5116-1GEA 190 EUR
APOSPC5116-1G APOSPC5116-5G
4-(2,2,2-Trifluoroethylcarbamoyl)phenylboronic acid 98%
4-(2,2,2-Trifluoroethylcarbamoyl)phenylboronic acid
Formula: C₉H₉BF₃NO₃
MW: 246,9800 g/mol
Melting Pt: 210…212 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08235087
CAS Number: 874459-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR