2-Chloro-3-fluorobenzotrifluoride
Supplier: Apollo Scientific
|
Warning
|
PC51197-1G
PC51197-5G
PC51197-25G
APOSPC51197-1GEA
114
EUR
APOSPC51197-1G
APOSPC51197-5G
APOSPC51197-25G
2-Chloro-3-fluorobenzotrifluoride
2-Chloro-3-fluorobenzotrifluoride
Formula:
C₇H₃ClF₄ MW: 198,55 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11100537 CAS Number: 1099597-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...