2,3,4,5,6-Pentafluorotoluene 99%

Supplier: Apollo Scientific
Warning

PC5120-100G PC5120-25G PC5120-5G PC5120-250G
APOSPC5120-100GEA 782 EUR
APOSPC5120-100G APOSPC5120-25G APOSPC5120-5G APOSPC5120-250G
2,3,4,5,6-Pentafluorotoluene 99%
2,3,4,5,6-Pentafluorotoluene
Formula: C₇H₃F₅
MW: 182,09 g/mol
Boiling Pt: 116…118 °C
Melting Pt: –30 °C
Density: 1,439 g/cm³ (25 °C)
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00000298
CAS Number: 771-56-2
EINECS: 212-233-7
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR