2-Fluoro-3-hydroxy-6-nitrophenylacetone
Supplier: Apollo Scientific
Synonyms:
1-(2-Fluoro-3-hydroxy-6-nitrophenyl)propan-2-one
PC51375-1G
PC51375-250MG
APOSPC51375-1GEA
145
EUR
APOSPC51375-1G
APOSPC51375-250MG
2-Fluoro-3-hydroxy-6-nitrophenylacetone
2-Fluoro-3-hydroxy-6-nitrophenylacetone
Formula:
C₉H₈FNO₄ MW: 213,164 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11869874 CAS Number: 649736-31-2 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...