Methyl perfluorononanoate 98%

Supplier: Apollo Scientific

PC5139-100G PC5139-25G
APOSPC5139-100GEA 594 EUR
APOSPC5139-100G APOSPC5139-25G
Methyl perfluorononanoate 98%
Methyl perfluorononanoate
Formula: C₁₀H₃F₁₇O₂
Boiling Pt: 63…65 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00153213
CAS Number: 51502-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR