Pentafluoro(2,2,2-trifluoroethoxy)benzene

Supplier: Apollo Scientific
Warning

Synonyms: (2,2,2-Trifluoroethoxy)pentafluorobenzene

PC5148-25G PC5148-5G PC5148-1G
APOSPC5148-25GEA 219 EUR
APOSPC5148-25G APOSPC5148-5G APOSPC5148-1G
Pentafluoro(2,2,2-trifluoroethoxy)benzene
Pentafluoro(2,2,2-trifluoroethoxy)benzene
Formula: C₈H₂F₈O
MW: 266,09 g/mol
Boiling Pt: 144…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00155915
CAS Number: 6669-03-0
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR