Methyl-4,4,4-trifluoroacetoacetate 95%
Supplier: Apollo Scientific
|
Warning
|
PC5165-5G
APOSPC5165-5GEA
68.3
EUR
APOSPC5165-5G
Methyl-4,4,4-trifluoroacetoacetate 95%
Methyl 4,4,4-trifluoroacetoacetate
Formula:
C₅H₅F₃O₃ MW: 170,09 g/mol Boiling Pt: 116…118 °C Density: 1,34 g/cm³ (25 °C) Flash Pt: 40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041004 CAS Number: 83643-84-9 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...