2-Fluoro-5-iodophenol 97%

Supplier: Apollo Scientific
Warning

PC52001-1G PC52001-250MG
APOSPC52001-1GEA 193 EUR
APOSPC52001-1G APOSPC52001-250MG
2-Fluoro-5-iodophenol 97%
2-Fluoro-5-iodophenol
Formula: C₆H₄FIO
MW: 238,0000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16659616
CAS Number: 186589-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR