3-Chloro-2-fluorobenzotrifluoride
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Chloro-2-fluoro-3-(trifluoromethyl)benzene
PC52043-1G
PC52043-5G
APOSPC52043-1GEA
125
EUR
APOSPC52043-1G
APOSPC52043-5G
3-Chloro-2-fluorobenzotrifluoride
3-Chloro-2-fluorobenzotrifluoride
Formula:
C₇H₃ClF₄ MW: 198,5500 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11100532 CAS Number: 1099597-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...