3-Fluoro-4-methoxybenzenesulphonamide 98%
Supplier: Apollo Scientific
PC520518-10G
PC520518-1G
PC520518-5G
APOSPC520518-10GEA
792
EUR
APOSPC520518-10G
APOSPC520518-1G
APOSPC520518-5G
3-Fluoro-4-methoxybenzenesulphonamide 98%
3-Fluoro-4-methoxybenzenesulphonamide
Formula:
C₇H₈FNO₃S MW: 205,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02091381 CAS Number: 874767-60-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...