4-Hydroxy-3-(trifluoromethoxy)benzaldehyde 98%
Supplier: Apollo Scientific
|
Warning
|
PC520535-1G
PC520535-2.5G
APOSPC520535-1GEA
176
EUR
APOSPC520535-1G
APOSPC520535-2.5G
4-Hydroxy-3-(trifluoromethoxy)benzaldehyde 98%
4-Hydroxy-3-(trifluoromethoxy)benzaldehyde
Formula:
C₈H₅F₃O₃ MW: 206,12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04973780 CAS Number: 53104-95-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...