3-Methyl-2-nitrobenzotrifluoride 97%

Supplier: Apollo Scientific

Synonyms: 1-Methyl-2-nitro-3-(trifluoromethyl)benzene

PC520536-10G PC520536-25G PC520536-5G
APOSPC520536-10GEA 582 EUR
APOSPC520536-10G APOSPC520536-25G APOSPC520536-5G
3-Methyl-2-nitrobenzotrifluoride 97%
3-Methyl-2-nitrobenzotrifluoride
Formula: C₈H₆F₃NO₂
Storage Temperature: Ambient
CAS Number: 92891-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR