5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid 95%

Supplier: Apollo Scientific

Synonyms: 5-Bromo-2-(trifluoromethyl)isonicotinic acid

PC520588-1G PC520588-5G PC520588-25G
APOSPC520588-1GEA 48 EUR
APOSPC520588-1G APOSPC520588-5G APOSPC520588-25G
5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid 95%
5-Bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid
Formula: C₇H₃BrF₃NO₂
MW: 270,01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10687236
CAS Number: 749875-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR