4-Fluoro-1H-pyrazole 98%

Supplier: Apollo Scientific
Danger

PC520591-1G PC520591-250MG PC520591-5G
APOSPC520591-1GEA 23.2 EUR
APOSPC520591-1G APOSPC520591-250MG APOSPC520591-5G
4-Fluoro-1H-pyrazole 98%
4-Fluoro-1H-pyrazole
Formula: C₃H₃FN₂
Melting Pt: 38…40 °C
Storage Temperature: Ambient
MDL Number: MFCD10699125
CAS Number: 35277-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR