4-Ethylbenzotrifluoride 97%

Supplier: Apollo Scientific

PC520611-10G PC520611-1G PC520611-5G
APOSPC520611-10GEA 0 EUR
APOSPC520611-10G APOSPC520611-1G APOSPC520611-5G
4-Ethylbenzotrifluoride 97%
4-Ethylbenzotrifluoride
Formula: C₉H₉F₃
Storage Temperature: Ambient
MDL Number: MFCD11100540
CAS Number: 27190-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR