(3-Fluoropyridin-4-yl)methanol 99%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Fluoro-4-(hydroxymethyl)pyridine

PC520662-1G PC520662-5G
APOSPC520662-1GEA 21.9 EUR
APOSPC520662-1G APOSPC520662-5G
(3-Fluoropyridin-4-yl)methanol 99%
(3-Fluoropyridin-4-yl)methanol
Formula: C₆H₆FNO
MW: 127,12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11656367
CAS Number: 870063-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR