3-Bromo-5-(difluoromethyl)pyridine 95%

Supplier: Apollo Scientific

PC520723-500MG PC520723-5G PC520723-1G
APOSPC520723-500MGEA 114 EUR
APOSPC520723-500MG APOSPC520723-5G APOSPC520723-1G
3-Bromo-5-(difluoromethyl)pyridine 95%
3-Bromo-5-(difluoromethyl)pyridine
Formula: C₆H₄BrF₂N
MW: 208.006 g/mol
Boiling Pt: 35…40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16140196
CAS Number: 114468-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR