2,6-Dichloro-4-(difluoromethyl)pyridine 97%
Supplier: Apollo Scientific
PC520728-1G
PC520728-5G
APOSPC520728-1GEA
396
EUR
APOSPC520728-1G
APOSPC520728-5G
2,6-Dichloro-4-(difluoromethyl)pyridine 97%
2,6-Dichloro-4-(difluoromethyl)pyridine
Formula:
C₆H₃Cl₂F₂N Storage Temperature: Ambient |
MDL Number:
MFCD16658727 CAS Number: 1201675-01-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...