3-Bromo-4-(difluoromethyl)pyridine 95%

Supplier: Apollo Scientific

PC520738-1G PC520738-5G
APOSPC520738-1GEA 350 EUR
APOSPC520738-1G APOSPC520738-5G
3-Bromo-4-(difluoromethyl)pyridine 95%
3-Bromo-4-(difluoromethyl)pyridine
Formula: C₆H₄BrF₂N
Storage Temperature: Ambient
MDL Number: MFCD18257232
CAS Number: 114468-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR