3-Benzyloxy-5-(trifluoromethyl)pyridine 97%

Supplier: Apollo Scientific

PC520741-10G PC520741-1G PC520741-5G
APOSPC520741-10GEA 1160 EUR
APOSPC520741-10G APOSPC520741-1G APOSPC520741-5G
3-Benzyloxy-5-(trifluoromethyl)pyridine 97%
3-Benzyloxy-5-(trifluoromethyl)pyridine
Formula: C₁₃H₁₀F₃NO
MW: 253,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18384832
CAS Number: 1256835-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR