4,4-Difluoropiperidine-1-sulphonyl chloride 95%

Supplier: Apollo Scientific

PC520746-250MG PC520746-500MG
APOSPC520746-250MGEA 792 EUR
APOSPC520746-250MG APOSPC520746-500MG
4,4-Difluoropiperidine-1-sulphonyl chloride 95%
4,4-Difluoropiperidine-1-sulphonyl chloride
Formula: C₅H₈ClF₂NO₂S
MW: 219,64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18917106
CAS Number: 1224929-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR