2-Chloro-3-(difluoromethyl)-5-fluoropyridine 95%

Supplier: Apollo Scientific

PC520759-250MG PC520759-500MG
APOSPC520759-250MGEA 558 EUR
APOSPC520759-250MG APOSPC520759-500MG
2-Chloro-3-(difluoromethyl)-5-fluoropyridine 95%
2-Chloro-3-(difluoromethyl)-5-fluoropyridine
Formula: C₆H₃ClF₃N
Storage Temperature: Ambient
MDL Number: MFCD22199309
CAS Number: 1374659-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR