2-Chloro-3-methylbenzotrifluoride 95%

Supplier: Apollo Scientific

PC520793-1G PC520793-5G
APOSPC520793-1GEA 187 EUR
APOSPC520793-1G APOSPC520793-5G
2-Chloro-3-methylbenzotrifluoride 95%
2-Chloro-3-methylbenzotrifluoride
Formula: C₈H₆ClF₃
Storage Temperature: Ambient
MDL Number: MFCD11226661
CAS Number: 74483-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR