4-(Trifluoromethyl)cyclohexylamine 97%

Supplier: Apollo Scientific

Synonyms: 4-(Trifluoromethyl)cyclohexanamine

PC520880-1G PC520880-5G
APOSPC520880-1GEA 277 EUR
APOSPC520880-1G APOSPC520880-5G
4-(Trifluoromethyl)cyclohexylamine 97%
4-(Trifluoromethyl)cyclohexylamine
Formula: C₇H₁₂F₃N
MW: 167,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09040573
CAS Number: 58665-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR