α,α,α-Trifluoro-o-toluoyl chloride 97%

Supplier: Apollo Scientific

Synonyms: 2-(Trifluoromethyl)benzoyl chloride, 2-(Trifluoromethyl)benzene-1-carbonyl chloride

PC52237-100G PC52237-500G PC52237-1KG
APOSPC52237-100GEA 56.7 EUR
APOSPC52237-100G APOSPC52237-500G APOSPC52237-1KG
α,α,α-Trifluoro-o-toluoyl chloride 97%
α,α,α-Trifluoro-o-toluoyl chloride
Formula: C₈H₄ClF₃O
Boiling Pt: 84…85 °C/16mmHg
Melting Pt: –22 °C
Density: 1,416 g/cm³ (25 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00000667
CAS Number: 312-94-7
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR