3-Bromo-6-chloro-2-fluorobenzaldehyde
Supplier: Apollo Scientific
|
Warning
|
PC53021-1G
PC53021-5G
PC53021-10G
PC53021-25G
APOSPC53021-1GEA
42.3
EUR
APOSPC53021-1G
APOSPC53021-5G
APOSPC53021-10G
APOSPC53021-25G
3-Bromo-6-chloro-2-fluorobenzaldehyde
3-Bromo-6-chloro-2-fluorobenzaldehyde
Formula:
C₇H₃BrClFO MW: 237,4500 g/mol Melting Pt: 92…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD11111905 CAS Number: 886615-30-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...