3-(Trifluoromethyl)benzoic acid hydrazide 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-(Trifluoromethyl)benzhydrazide, 3-(Trifluoromethyl)benzohydrazide
PC5311-10G
PC5311-1G
PC5311-5G
APOSPC5311-10GEA
164
EUR
APOSPC5311-10G
APOSPC5311-1G
APOSPC5311-5G
3-(Trifluoromethyl)benzoic acid hydrazide 95%
3-(Trifluoromethyl)benzoic acid hydrazide
Formula:
C₈H₇F₃N₂O MW: 204,1492 g/mol Melting Pt: 113…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00221476 CAS Number: 22227-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...