1-Fluorononane 98%

Supplier: Apollo Scientific
Danger

PC5314-1G PC5314-5G
APOSPC5314-1GEA 0 EUR
APOSPC5314-1G APOSPC5314-5G
1-Fluorononane 98%
1-Fluorononane
Formula: C₉H₁₉F
Boiling Pt: 166…169 °C/760mmHg
Density: 0,81 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000455
CAS Number: 463-18-3
UN: 2929
ADR: 6.1+3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR