Diethyl (4-fluorobenzyl)phosphonate 98%

Supplier: Apollo Scientific
Warning

Synonyms: (4-Fluorobenzyl)phosphonic acid diethyl ester, Diethyl 4-fluorobenzylphosphonate

PC53237-25G PC53237-5G
APOSPC53237-25GEA 275 EUR
APOSPC53237-25G APOSPC53237-5G
Diethyl (4-fluorobenzyl)phosphonate 98%
Diethyl (4-fluorobenzyl)phosphonate
Formula: C₁₁H₁₆FO₃P
MW: 246,2200 g/mol
Boiling Pt: 150 °C/6mmHg
Density: 1,1425 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD06795966
CAS Number: 63909-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR