3-Fluoro-4-nitropyridine-1-oxide 98%

Supplier: Apollo Scientific
Warning

PC53338-25G PC53338-5G PC53338-100G
APOSPC53338-25GEA 127 EUR
APOSPC53338-25G APOSPC53338-5G APOSPC53338-100G
3-Fluoro-4-nitropyridine-1-oxide 98%
3-Fluoro-4-nitropyridine-1-oxide
Formula: C5H3FN2O3
Storage Temperature: Ambient
MDL Number: MFCD00234027
CAS Number: 769-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR