3-Fluoro-5-nitrotoluene
Supplier: Apollo Scientific
|
Warning
|
PC53458-25G
PC53458-5G
PC53458-100G
APOSPC53458-25GEA
46.4
EUR
APOSPC53458-25G
APOSPC53458-5G
APOSPC53458-100G
3-Fluoro-5-nitrotoluene
3-Fluoro-5-nitrotoluene
Formula:
C7H6FNO2 Storage Temperature: Ambient |
MDL Number:
MFCD06797943 CAS Number: 499-08-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...