2-Bromo-5-(trifluoromethyl)benzenesulphonamide

Supplier: Apollo Scientific
Warning

PC5411-1G PC5411-5G
APOSPC5411-1GEA 34.8 EUR
APOSPC5411-1G APOSPC5411-5G
2-Bromo-5-(trifluoromethyl)benzenesulphonamide
2-Bromo-5-(trifluoromethyl)benzenesulphonamide
Formula: C₇H₅BrF₃NO₂S
MW: 304,09 g/mol
Melting Pt: 185…189 °C
Storage Temperature: Ambient
CAS Number: 351003-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR