Pentafluorobenzoic acid 99%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2,3,4,5,6-Pentafluorobenzoic acid
PC5500-100G
PC5500-1KG
PC5500-25G
APOSPC5500-100GEA
31.9
EUR
APOSPC5500-100G
APOSPC5500-1KG
APOSPC5500-25G
Pentafluorobenzoic acid 99%
Pentafluorobenzoic acid
Formula:
C₇HF₅O₂ MW: 212.08 g/mol Boiling Pt: 219…221 °C Melting Pt: 100…104 °C Density: 1.944 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002406 CAS Number: 602-94-8 EINECS: 210-026-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...